missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Invitrogen™ Total Exosome RNA & Protein Isolation Kit
The Total Exosome RNA and Protein Isolation Kit enables isolation of total RNA and proteins from exosome samples extracted from biological fluids (e.g., by using a Total Exosome Isolation (from cell culture media) or Total Exosome Isolation (from serum).
Supplier: Invitrogen™ 4478545
Description
The Total Exosome RNA and Protein Isolation Kit enables isolation of total RNA and proteins from exosome samples extracted from biological fluids (e.g., by using a Total Exosome Isolation (from cell culture media) or Total Exosome Isolation (from serum).
- Extracts highly pure total RNA (including small RNA fraction) from exosomes
- Recovers protein and RNA from the same sample
- Compatible with all protocols for exosome isolation
Not high-throughput compatible.
Order Info
Shipping Condition: Room Temperature
Specifications
Store at 2°C to 8°C. | |
Total Exosome Isolation | |
Not High-throughput Compatible (Manual) | |
Total RNA, Protein, RNA & Protein, miRNA | |
Organelle & Vesicle-Related Samples, Exosomes |
Next-Generation Sequencing, Real-Time Quantitative PCR (qPCR), Western Blot, microRNA Analysis | |
Total RNA, Protein, micro RNA | |
Spin Column (Glass Fiber Filter), Organic Extraction | |
40 preps | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.