missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ UltraPure™ 1M Tris-HCI, pH 8.0 DFS Item

Pre-mixed, pH-adjusted, sterile-filtered solutions

Supplier:  Invitrogen™ 15568025

Catalog No. 15568025


Only null left
Explore available promotions

Description

Description

  • Prepared as 1M concentrate, buffers can be diluted to desired concentration
  • Used in molecular biology or general biochemistry applications
  • Performance and Quality Testing: No DNase, RNase, or protease activity detected
  • Sustainable packaging

Agarose Gel Electrophoresis, DNA & RNA Purification & Analysis, DNA Extraction, General gDNA Purification Reagents & Accessories, Genomic DNA Purification, Maxiprep, Midiprep, Miniprep, Nucleic Acid Gel Electrophoresis & Blotting, Plasmid DNA Purification

Order Info

Shipping Condition: Room Temperature

Specifications

Specifications

1 M
pH 8.0
Tris-HCl Buffer
Room Temperature
Store in refrigerator (2–8°C).
UltraPure
1 L
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.