missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ UltraPure™ Acrylamide DFS Item

Recrystallized three times to ensure highly purified product

Supplier:  Invitrogen™ 15512023

Catalog No. 15512023


Explore available promotions

Description

Description

UltraPure™ Acrylamide is major ingredient in polyacrylamide gels, which are used for electrophoretic separation of both nucleic acid fragments and proteins. DNA fragments generated by restriction endonuclease digestion, dideoxy sequencing, or Maxam and Gilbert sequencing methods can be separated by polyacrylamide gel electrophoresis (PAGE).

  • Proteins can be separated by variety of techniques with polyacrylamide gels, including denaturing gels using SDS or urea, isoelectric focusing gels, and native gels in a wide variety of buffers
  • Performance and Quality Testing: Polymerization time is evaluated in an SDS polyacrylamide gel system

Acrylamide Gel Electrophoresis, DNA & RNA Purification & Analysis, Gel Casting, Nucleic Acid Gel Electrophoresis & Blotting, Protein Gel Electrophoresis, Proteins, Expression, Isolation & Analysis

Order Info

Shipping Condition: Room Temperature

Specifications

Specifications

Store at room temperature.
Gel Electrophoresis
500 g
Acrylamide
UltraPure™
Room Temperature
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.