Invitrogen™ UltraPure™ TBE Buffer, 10X DFS Item
Compatible with agarose gels
Supplier:  Invitrogen™ 15581028
 View more versions of this product
Catalog No. 15581028
Explore available promotions
Description
Description
UltraPure 10X TBE Buffer is a sterile-filtered solution of 1M Tris, 0.9 M boric acid, and 0.01M EDTA used to prepare 1X buffer for polyacrylamide and agarose gel electrophoresis.
  • UltraPure 10X TBE Buffer is available in 1 L plastic bottle/10 L Cubitainer™ box
  • Performance and quality testing: No DNase, RNase, or protease activity detected. Sustainable packaging
Acrylamide Gel Electrophoresis, DNA & RNA Purification & Analysis, Nucleic Acid Gel Electrophoresis & Blotting
Specifications
Specifications
Running Buffer
Store at room temperature.
Polyacrylamide Gels, Agarose Gels
UltraPure™
10 L
10 X
Bottle
Nucleic Acid Gel Electrophoresis, Blotting
TBE Buffer
Room Temperature
Videos
SDS
Documents
Documents
Promotions
Promotions
For Research Use Only. Not for use in diagnostic procedures.