missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Invitrogen™ UltraPure™ TBE Buffer, 10X
Compatible with agarose gels
Supplier: Invitrogen™ 15581044
Description
UltraPure 10X TBE Buffer is a sterile-filtered solution of 1M Tris, 0.9 M boric acid, and 0.01M EDTA used to prepare 1X buffer for polyacrylamide and agarose gel electrophoresis.
- UltraPure 10X TBE Buffer is available in 1 L plastic bottle/10 L Cubitainer™ box
- Performance and quality testing: No DNase, RNase, or protease activity detected. Sustainable packaging
Acrylamide Gel Electrophoresis, DNA & RNA Purification & Analysis, Nucleic Acid Gel Electrophoresis & Blotting
Specifications
Running Buffer | |
Contents: One bottle (1 L) UltraPure™ 10X TBE Buffer Storage: Room Temperature |
|
Polyacrylamide Gels, Agarose Gels | |
UltraPure™ | |
1 L |
10 X | |
Bottle | |
Nucleic Acid Gel Electrophoresis, Blotting | |
TBE Buffer | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.