Invitrogen™ X-Gal DFS Item
X-Gal (5-bromo-4-chloro-3-indolyl-β-D-galactoside) is a histochemical substrate for β-galactosidase. This substrate yields a blue precipitate upon hydrolysis, making it suitable for use in immunoblotting or immunocytochemical assays.
Supplier:  Invitrogen™ 15520018
 View more versions of this product
Catalog No. 15520018
Explore available promotions
Description
Description
X-Gal (5-bromo-4-chloro-3-indolyl-β-D-galactoside) is a histochemical substrate for β-galactosidase. This substrate yields a blue precipitate upon hydrolysis, making it suitable for use in immunoblotting or immunocytochemical assays. Bacteria containing active β-galactosidase produce blue colonies when grown on plates containing X-Gal. X-Gal is supplied as white crystals and is soluble in dimethylformamide.
Primary applications
  • Blue-white screening—detect functional β-galactosidase (reporter gene expression)
  • Histochemical detection of β-galactosidase activity in fixed tissues
  • X-gal overlay assay
Find the media format that fits your needs
We offer many ready-to-use media formulations, such as LB Broth (10855001), Terrific Broth (A1374301) or S.O.C recovery medium (15544034).
We also offer IPTG (15529019) for blue-white screening.
Explore other media formats for bacterial growth.
Specifications
Specifications
5-bromo-4-chloro-3-indolyl-β-D-galactoside
Colorimetric
Bottle
X-Gal Substrate
Approved for shipment at Room Temperature or on Wet Ice
• X-Gal, powder, 1 g
X-Gal, Cell-Permeant
Immunoblotting
1 g
Powder
Videos
SDS
Documents
Documents
Promotions
Promotions
For Research Use Only. Not for use in diagnostic procedures.