missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Iodic acid, 99.6%, Thermo Scientific Chemicals

$381.75 - $381.75

Chemical Identifiers

CAS 7782-68-5
Molecular Formula ILiO3
Molecular Weight (g/mol) 181.84
MDL Number MFCD00011348
InChI Key FZAXZVHFYFGNBX-UHFFFAOYSA-M
Synonym unii-6u8j18jsbm, 6u8j18jsbm, iodicacid, monoiodic acid, caswell no. 499a, iodsaeure, trioxoiodic acid, iodic v acid, sodium iodate min
PubChem CID 24345
ChEBI CHEBI:24857
IUPAC Name lithium(1+) iodate
SMILES [Li+].[O-][I](=O)=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC423811000
SDS View Documents
Thermo Scientific Chemicals
423811000
100 g Glass bottle
Each for $381.75
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
Specifications

Chemical Identifiers

7782-68-5
181.84
FZAXZVHFYFGNBX-UHFFFAOYSA-M
24345
lithium(1+) iodate
ILiO3
MFCD00011348
unii-6u8j18jsbm, 6u8j18jsbm, iodicacid, monoiodic acid, caswell no. 499a, iodsaeure, trioxoiodic acid, iodic v acid, sodium iodate min
CHEBI:24857
[Li+].[O-][I](=O)=O

Specifications

White
7782-68-5
10ppm max.
0.02% max.
99.6%
ILiO3
HIO3
15, 5056
Solubility in water: 2690g/l (20°C). Other solubilities: soluble in nitric acid, soluble in diluted alcohol, insoluble in absolute alcohol, ether and, chloroform
[Li+].[O-][I](=O)=O
181.84
CHEBI:24857
99.6%
Iodic acid, For analysis ACS
110.0°C
4.6200g/mL
Glass bottle
4.62
100 g
Crystalline Powder
MFCD00011348
unii-6u8j18jsbm, 6u8j18jsbm, iodicacid, monoiodic acid, caswell no. 499a, iodsaeure, trioxoiodic acid, iodic v acid, sodium iodate min
FZAXZVHFYFGNBX-UHFFFAOYSA-M
lithium(1+) iodate
24345
175.91
ACS Reagent
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
May intensify fire; oxidizer.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only