missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Iodic acid, ACS, 99.5% min, Thermo Scientific Chemicals

$171.12 - $597.16

Chemical Identifiers

CAS 7782-68-5
Molecular Formula ILiO3
Molecular Weight (g/mol) 181.84
MDL Number MFCD00011348
InChI Key FZAXZVHFYFGNBX-UHFFFAOYSA-M
Synonym unii-6u8j18jsbm, 6u8j18jsbm, iodicacid, monoiodic acid, caswell no. 499a, iodsaeure, trioxoiodic acid, iodic v acid, sodium iodate min
PubChem CID 24345
ChEBI CHEBI:24857
IUPAC Name lithium(1+) iodate
SMILES [Li+].[O-][I](=O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA8768118
View Documents
Thermo Scientific Chemicals
08768118
50 g N/A
 
AA8768130
View Documents
Thermo Scientific Chemicals
08768130
250 g N/A
 
Description

Description

Iodic acid is used in analytical chemistry laboratories to standardize solutions of both weak and strong bases using methyl red or methyl orange as the indicator. It acts as a key starting material to synthesize sodium or potassium iodate, thereby increasing the iodine content of salt.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7782-68-5
181.84
FZAXZVHFYFGNBX-UHFFFAOYSA-M
24345
lithium(1+) iodate
ILiO3
MFCD00011348
unii-6u8j18jsbm, 6u8j18jsbm, iodicacid, monoiodic acid, caswell no. 499a, iodsaeure, trioxoiodic acid, iodic v acid, sodium iodate min
CHEBI:24857
[Li+].[O-][I](=O)=O

Specifications

Powder/chunks
7782-68-5
Light sensitive
50 g
Powder/Chunks
unii-6u8j18jsbm, 6u8j18jsbm, iodicacid, monoiodic acid, caswell no. 499a, iodsaeure, trioxoiodic acid, iodic v acid, sodium iodate min
FZAXZVHFYFGNBX-UHFFFAOYSA-M
lithium(1+) iodate
24345
175.91
Iodic Acid
110°C (decomposition)
4.629 g/cm3
99.5%
ILiO3
MFCD00011348
Soluble in water and methanol.
[Li+].[O-][I](=O)=O
181.84
CHEBI:24857
ACS
Videos
Safety and Handling

Safety and Handling

P210-P220-P221-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H272-H314-H335

DOTInformation : Hazard Class: 5.1; Packaging Group: II

EINECSNumber : 231-962-1

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only