missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Iodine, Certified, 0.1000N ±0.0005N (0.05M), LabChem™
Supplier: LabChem LC156604
Specifications
Iodine, 0.1N (0.05M) | |
96.17,2.54,1.27,0.02 | |
4 L | |
I2 | |
water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide | |
PNDPGZBMCMUPRI-UHFFFAOYSA-N | |
diiodine | |
253.809 | |
Amber Glass |
7553-56-2,7681-11-0,7681-11-0,7647-01-0 | |
Liquid | |
I2 | |
MFCD00011355 MFCD00164163 | |
Soluble in water | |
II | |
253.81 | |
Certified |
Chemical Identifiers
7553-56-2 | |
253.81 | |
PNDPGZBMCMUPRI-UHFFFAOYSA-N | |
II |
I2 | |
MFCD00011355 MFCD00164163 | |
diiodine |
Safety and Handling
GHS H Statement
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.
Harmful to aquatic life.
GHS P Statement
Avoid breathing mist, vapors, spray.
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
Contaminated work clothing must not be allowed out of the workplace.
Avoid release to the e
Warning