missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iodine (Iodine-Iodide), 0.0200 N (N/50), Ricca Chemical
Supplier: Ricca Chemical Company 39731
Specifications
| Iodine (Iodine-Iodide), Ricca Chemical | |
| 253.81 g/mol | |
| 0.02N | |
| 4 L |
| 7732-18-5 | |
| Laboratory | |
| Amber Glass Bottle |
Chemical Identifiers
| 0.0 |