Learn More
Iodine monobromide, 98%, Thermo Scientific Chemicals
$80.14 - $452.99
Chemical Identifiers
CAS | 7789-33-5 |
---|---|
Molecular Formula | BrI |
Molecular Weight (g/mol) | 206.81 |
MDL Number | MFCD00011353 |
InChI Key | CBEQRNSPHCCXSH-UHFFFAOYSA-N |
Synonym | iodine monobromide, iodine bromide, iodobromine, iodine bromide ibr, unii-g0k622rd9n, iodinemonobromide, iodobromane, bromoiodine, jodmonobromid, hanus solution |
PubChem CID | 82238 |
IUPAC Name | iodobromane |
SMILES | BrI |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1615814
|
Thermo Scientific Chemicals
A1615814 |
25 g |
N/A
|
|
|||||
AAA1615822
|
Thermo Scientific Chemicals
A1615822 |
100 g |
N/A
|
|
|||||
AAA1615830
|
Thermo Scientific Chemicals
A1615830 |
250 g |
N/A
|
|
|||||
Description
Iodine monobromide is used as an electrophile employed in a new synthetic approach to polyketide structural units. It is also used in iodometry and serves as a source of I+. It is a powerful iodinating agent used in organic synthesis as well as involved in the synthesis of radioiodinated fatty acids for heart imaging.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIodine monobromide is used as an electrophile employed in a new synthetic approach to polyketide structural units. It is also used in iodometry and serves as a source of I+. It is a powerful iodinating agent used in organic synthesis as well as involved in the synthesis of radioiodinated fatty acids for heart imaging.
Solubility
Soluble in water, alcohol, ether, carbon disulfide and glacial acetic acid.
Notes
Air, light and moisture sensitive. Store in cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with alcohols, phosphorus, sodium and sodiumoxides and potassium.
Chemical Identifiers
7789-33-5 | |
206.81 | |
CBEQRNSPHCCXSH-UHFFFAOYSA-N | |
82238 | |
BrI |
BrI | |
MFCD00011353 | |
iodine monobromide, iodine bromide, iodobromine, iodine bromide ibr, unii-g0k622rd9n, iodinemonobromide, iodobromane, bromoiodine, jodmonobromid, hanus solution | |
iodobromane |
Specifications
116°C (decomposition) | |
7789-33-5 | |
BrI | |
UN3260 | |
None | |
iodine monobromide, iodine bromide, iodobromine, iodine bromide ibr, unii-g0k622rd9n, iodinemonobromide, iodobromane, bromoiodine, jodmonobromid, hanus solution | |
CBEQRNSPHCCXSH-UHFFFAOYSA-N | |
iodobromane | |
82238 | |
98% | |
Light and moisture sensitive | |
Iodine monobromide |
50°C (sublimation) | |
25 g | |
MFCD00011353 | |
3902973 | |
14,5016 | |
Soluble in water,alcohol,ether,carbon disulfide and glacial acetic acid. | |
BrI | |
206.81 | |
206.81 | |
Pungent | |
4.416 |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
EINECSNumber : 232-159-9
TSCA : Yes
Recommended Storage : Keep cold; Store under Nitrogen
RUO – Research Use Only