missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Iodine monochloride, approx. 0.22N soln. in glacial acetic acid, Thermo Scientific Chemicals

For Determination of Iodine Absorption Number of Fats

Supplier:  Thermo Scientific Chemicals 042647AP

 View more versions of this product

Catalog No. AA42647AP


Explore available promotions

Description

Description

For determination of iodine absorption number of fatsIodine monochloride is used as a catalyst in organic synthesis. It is the source of electrophilic iodine in the synthesis of certain aromatic iodides. It is used to determine the iodine value of a substance.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Moisture and Light Sensitive
Specifications

Specifications

Iodine monochloride
Liquid
ClI
iodine monochloride, iodine chloride, iodochlorine, iodine chloride icl, chloroiodide, wijs' chloride, iodinemonochloride, chlorine iodide, unii-0smg5nlu45, protochlorure d'iode french
QZRGKCOWNLSUDK-UHFFFAOYSA-N
iodochlorane
24640
485°C (905°F) (acetic acid)
Liquid
7790-99-0
500 mL
MFCD00011354
Miscible with water,alcohol,ether and carbon disulfide.
ClI
162.35
162.36
Light and moisture sensitive

Chemical Identifiers

7790-99-0
162.35
QZRGKCOWNLSUDK-UHFFFAOYSA-N
24640
ClI
ClI
MFCD00011354
iodine monochloride, iodine chloride, iodochlorine, iodine chloride icl, chloroiodide, wijs' chloride, iodinemonochloride, chlorine iodide, unii-0smg5nlu45, protochlorure d'iode french
iodochlorane
Safety and Handling

Safety and Handling

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H226-H314-H335

DOTInformation : Hazard Class: 8, Packing Group: II

EINECSNumber : 232-236-7

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only