missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Iodine, resublimed crystals, ACS, 99.8%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 04195522

 View more versions of this product

Catalog No. AA4195522


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

183°C to 186° (sublimation)
7553-56-2
Prills or Crystals
99.8%
MFCD00011355 MFCD00164163
PNDPGZBMCMUPRI-UHFFFAOYSA-N
molecular iodine
807
253.81
Irritating
4.930 g/cm3
Iodine
Gray-Purple
100 g
I2
iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode
II
253.81
CHEBI:17606
ACS
Bluish-black

Chemical Identifiers

7553-56-2
253.81
PNDPGZBMCMUPRI-UHFFFAOYSA-N
807
molecular iodine
I2
MFCD00011355 MFCD00164163
iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode
CHEBI:17606
II
Videos
Safety and Handling

Safety and Handling

P260-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P314-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335-H372

DOTInformation : Hazard Class: 8; Packaging Group: III

EINECSNumber : 231-442-4

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only