Learn More
Iodoacetamide, 98%, Thermo Scientific Chemicals
$102.80 - $1077.73
Chemical Identifiers
CAS | 144-48-9 |
---|---|
Molecular Formula | C2H4INO |
Molecular Weight (g/mol) | 184.96 |
InChI Key | PGLTVOMIXTUURA-UHFFFAOYSA-N |
Synonym | iodoacetamide, monoiodoacetamide, surauto, acetamide, 2-iodo, 2-iodo-acetamide, usaf d-1, alpha-iodoacetamide, ccris 7710, unii-zrh8m27s79, n-3-methoxy-4-butoxybenzyl thiobarbituric acid |
PubChem CID | 3727 |
IUPAC Name | 2-iodoacetamide |
SMILES | C(C(=O)N)I |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC122270050
|
Thermo Scientific Chemicals
122270050 |
5 g |
Each for $102.80
|
|
|||||
AC122270250
|
Thermo Scientific Chemicals
122270250 |
25 g |
Each for $339.38
|
|
|||||
AC122271000
|
Thermo Scientific Chemicals
122271000 |
100 g |
Each for $1,077.73
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
144-48-9 | |
184.96 | |
iodoacetamide, monoiodoacetamide, surauto, acetamide, 2-iodo, 2-iodo-acetamide, usaf d-1, alpha-iodoacetamide, ccris 7710, unii-zrh8m27s79, n-3-methoxy-4-butoxybenzyl thiobarbituric acid | |
2-iodoacetamide |
C2H4INO | |
PGLTVOMIXTUURA-UHFFFAOYSA-N | |
3727 | |
C(C(=O)N)I |
Specifications
144-48-9 | |
Beige or White to Yellow | |
97.5% min. (ex I) (Argentometry) | |
C2H4INO | |
5 g | |
01,504 | |
Solubility in water: slightly soluble | |
C(C(=O)N)I | |
184.96 | |
184.96 | |
Crystals or Powder |
>90.0°C | |
Authentic | |
Glass bottle | |
ICH2CONH2 | |
02,223 | |
iodoacetamide, monoiodoacetamide, surauto, acetamide, 2-iodo, 2-iodo-acetamide, usaf d-1, alpha-iodoacetamide, ccris 7710, unii-zrh8m27s79, n-3-methoxy-4-butoxybenzyl thiobarbituric acid | |
PGLTVOMIXTUURA-UHFFFAOYSA-N | |
2-iodoacetamide | |
3727 | |
98% | |
Iodoacetamide |
Safety and Handling
GHS H Statement
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
Causes severe skin burns and eye damage.
Toxic if swallowed.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee
GHS Signal Word: Danger
EINECSNumber : 205-630-1
RUO – Research Use Only