missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Iodoacetic acid, sodium salt, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 170970250

 View more versions of this product

Catalog No. AC170970250


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Iodoacetic acid, sodium salt
208.0°C to 210.0°C
Authentic
Glass bottle
ICH2CO2Na
25 g
13,116
Solubility in water: soluble. Other solubilities: soluble in alcohol,slightly soluble in ether
C(C(=O)[O-])I.[Na+]
207.93
207.93
Powder
305-53-3
Brown-Yellow to Yellow
99%
C2H2INaO2
MFCD00002686
02, 222
sodium iodoacetate, iodoacetic acid sodium salt, acetic acid, iodo-, sodium salt, sodium 2-iodoacetate, sodium monoiodoacetate, iodoacetate, sodium salt, sodium iodacetate, iodoacetic acid, sodium salt, unii-07814f2qri, ccris 9232
AGDSCTQQXMDDCV-UHFFFAOYSA-M
sodium;2-iodoacetate
5239
99%

Chemical Identifiers

305-53-3
207.93
AGDSCTQQXMDDCV-UHFFFAOYSA-M
5239
C(C(=O)[O-])I.[Na+]
C2H2INaO2
MFCD00002686
sodium iodoacetate, iodoacetic acid sodium salt, acetic acid, iodo-, sodium salt, sodium 2-iodoacetate, sodium monoiodoacetate, iodoacetate, sodium salt, sodium iodacetate, iodoacetic acid, sodium salt, unii-07814f2qri, ccris 9232
sodium;2-iodoacetate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye damage.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.

GHS Signal Word: Danger

EINECSNumber : 206-165-7

RTECSNumber : AI3675000

TSCA : TSCA

Recommended Storage : −20°C

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only