missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Iodoacetic anhydride, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 307592500

 View more versions of this product

Catalog No. AC307592500


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Iodoacetic anhydride
43.0°C to 49.0°C
>110°C
97%
C4H4I2O3
MFCD00001080
iodoacetic anhydride, di iodoacetic anhydride, acmc-20akq5, iodoacetic acid anhydride, 2-iodoacetyl 2-iodoacetate, acetic acid, iodo-, anhydride, acetic acid, 2-iodo-,1,1'-anhydride, iodoacetic anhydride, technical t
C(C(=O)OC(=O)CI)I
353.88
353.88
Crystalline Powder
54907-61-8
Yellow
Authentic
Glass bottle
(ICH2CO)2O
250 mg
RBNSZWOCWHGHMR-UHFFFAOYSA-N
(2-iodoacetyl) 2-iodoacetate
2733254
97%

Chemical Identifiers

54907-61-8
353.88
RBNSZWOCWHGHMR-UHFFFAOYSA-N
2733254
C(C(=O)OC(=O)CI)I
C4H4I2O3
MFCD00001080
iodoacetic anhydride, di iodoacetic anhydride, acmc-20akq5, iodoacetic acid anhydride, 2-iodoacetyl 2-iodoacetate, acetic acid, iodo-, anhydride, acetic acid, 2-iodo-,1,1'-anhydride, iodoacetic anhydride, technical t
(2-iodoacetyl) 2-iodoacetate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Toxic if inhaled.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove to fresh air

GHS Signal Word: Danger

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only