missing translation for 'onlineSavingsMsg'
Learn More

Iodoacetonitrile, 95%

CAS: 624-75-9 | C2H2IN | 166.95 g/mol

$303.51 - $303.51

Chemical Identifiers

CAS 624-75-9
Molecular Formula C2H2IN
Molecular Weight (g/mol) 166.95
MDL Number MFCD00001886
InChI Key VODKOOOHHCAWFR-UHFFFAOYSA-N
Synonym iodoacetonitrile, acetonitrile, iodo, iodo-acetonitrile, 2-iodoethanenitrile, 2-iodo acetonitrile, 2-iodoaceto-nitrile, 2-iodanylethanenitrile, pubchem12543, acmc-1ba3n
PubChem CID 69356
IUPAC Name 2-iodoacetonitrile
SMILES C(C#N)I
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC122290050
SDS View Documents
Thermo Scientific Chemicals
122290050
5 mL Glass bottle
Each for $303.51
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

624-75-9
166.95
VODKOOOHHCAWFR-UHFFFAOYSA-N
69356
C(C#N)I
C2H2IN
MFCD00001886
iodoacetonitrile, acetonitrile, iodo, iodo-acetonitrile, 2-iodoethanenitrile, 2-iodo acetonitrile, 2-iodoaceto-nitrile, 2-iodanylethanenitrile, pubchem12543, acmc-1ba3n
2-iodoacetonitrile

Specifications

624-75-9
2.3000g/mL
86°C
94% min. (GC)
C2H2IN
ICH2CN
5 mL
2.3
VODKOOOHHCAWFR-UHFFFAOYSA-N
2-iodoacetonitrile
69356
95%
Iodoacetonitrile
Brown to Red
182°C to 184°C (720.0 mmHg)
Authentic
Glass bottle
1.5740 to 1.578
MFCD00001886
02, 223
iodoacetonitrile, acetonitrile, iodo, iodo-acetonitrile, 2-iodoethanenitrile, 2-iodo acetonitrile, 2-iodoaceto-nitrile, 2-iodanylethanenitrile, pubchem12543, acmc-1ba3n
C(C#N)I
166.95
166.95
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Toxic if inhaled.
Toxic if swallowed.
Toxic in contact with skin.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 210-860-

SDS
Documents

Documents

RUO – Research Use Only