missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Iodoacetonitrile, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L0363409

 View more versions of this product

Catalog No. AAL0363409


Explore available promotions

Description

Description

Iodoacetonitrile is used for alkylation of nucleophiles. It undergoes Reformatsky and Wittig-type reactions with aldehydes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Iodoacetonitrile is used for alkylation of nucleophiles. It undergoes Reformatsky and Wittig-type reactions with aldehydes.

Solubility
Soluble in benzene, alcohol, acetone, ether. Slightly soluble in water. Insoluble in hexane.

Notes
Light Sensitive. Store at 4°C. Incompatible with reducing agents, light and bases. Store away from strong bases.
Specifications

Specifications

Iodoacetonitrile
2.306
86°C (186°F)
1.574
MFCD00001886
UN2922
Light sensitive
Soluble in benzene,alcohol,acetone,ether. Slightly soluble in water. Insoluble in hexane.
C(C#N)I
166.949
166.95
624-75-9
70°C to 72°C (10 mmHg)
C2H2IN
ICH2CN
10 g
1734628
iodoacetonitrile, acetonitrile, iodo, iodo-acetonitrile, 2-iodoethanenitrile, 2-iodo acetonitrile, 2-iodoaceto-nitrile, 2-iodanylethanenitrile, pubchem12543, acmc-1ba3n
VODKOOOHHCAWFR-UHFFFAOYSA-N
2-iodoacetonitrile
69356
97%

Chemical Identifiers

624-75-9
166.949
VODKOOOHHCAWFR-UHFFFAOYSA-N
69356
C(C#N)I
C2H2IN
MFCD00001886
iodoacetonitrile, acetonitrile, iodo, iodo-acetonitrile, 2-iodoethanenitrile, 2-iodo acetonitrile, 2-iodoaceto-nitrile, 2-iodanylethanenitrile, pubchem12543, acmc-1ba3n
2-iodoacetonitrile
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H302-H312-H332-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Combustible liquid.

P210-P235-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c

H227-H301+H311+H331-H314-H335

DOTInformation : Hazard Class: 6.1; Packaging Group: II

EINECSNumber : 210-860-0

RTECSNumber : AM0740000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only