Learn More
Iodocyclohexane, 98%, stab. with copper, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2484022
Description
Iodocyclohexane has been employed as reagent in demethylation of aryl methyl ethers in DMF under reflux condition.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIodocyclohexane has been employed as reagent in demethylation of aryl methyl ethers in DMF under reflux condition.
Solubility
Soluble in ethanol, ether, acetone.
Notes
Light sensitive. Store away from oxidizing agent, light.
Specifications
Iodocyclohexane | |
626-62-0 | |
180°C | |
C6H11I | |
MFCD00003826 | |
1900797 | |
cyclohexane, iodo, cyclohexyl iodide, cyclohexyliodide, iodo-cyclohexane, 1-iodocyclohexane, heptynyltrimethylsilane, iodocyclohexane, acmc-1att4, c-c6h11i | |
FUCOMWZKWIEKRK-UHFFFAOYSA-N | |
iodocyclohexane | |
12289 | |
98% |
stabilized with copper | |
1.625 | |
71°C (159°F) | |
1.5475 | |
100 g | |
Light sensitive | |
Soluble in ethanol,ether,acetone. | |
IC1CCCCC1 | |
210.06 | |
210.06 |
Chemical Identifiers
626-62-0 | |
210.06 | |
FUCOMWZKWIEKRK-UHFFFAOYSA-N | |
12289 | |
IC1CCCCC1 |
C6H11I | |
MFCD00003826 | |
cyclohexane, iodo, cyclohexyl iodide, cyclohexyliodide, iodo-cyclohexane, 1-iodocyclohexane, heptynyltrimethylsilane, iodocyclohexane, acmc-1att4, c-c6h11i | |
iodocyclohexane |
Safety and Handling
GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H227-H315-H319-H335
EINECSNumber : 210-957-8
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only