missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Iodomethane, 2M solution in tert-Butyl methyl ether, AcroSeal™, Thermo Scientific Chemicals

$193.72 - $193.72

Specifications

Chemical Name or Material Iodomethane
Name Note 2M Solution in tert-Butyl Ether
CAS 74-88-4
Density 0.9330g/mL
Boiling Point 41.0°C to 43.0°C
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC378411000
View Documents
Thermo Scientific Chemicals
378411000
100 mL AcroSeal™ Glass bottle
Each for $193.72
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Alkylation reagent
Specifications

Specifications

Iodomethane
74-88-4
41.0°C to 43.0°C
MFCD00001073
0.933
141.94
2M Solution in tert-Butyl Ether
0.9330g/mL
−18°C
15, 6159
141.94
SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only