missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Iodomethane, 99%, stabilized, Thermo Scientific Chemicals
$63.66 - $1443.96
Chemical Identifiers
CAS | 74-88-4 |
---|---|
Molecular Formula | CH3I |
Molecular Weight (g/mol) | 141.94 |
MDL Number | MFCD00001073 |
InChI Key | INQOMBQAUSQDDS-UHFFFAOYSA-N |
Synonym | methyl iodide, monoiodomethane, methane, iodo, methyljodid, jod-methan, methyliodide, methyljodide, iodometano, joodmethaan, metylu jodek |
PubChem CID | 6328 |
ChEBI | CHEBI:39282 |
IUPAC Name | iodomethane |
SMILES | CI |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC122370050
|
Thermo Scientific Chemicals
122370050 |
5 g | Glass bottle |
Each for $63.66
|
|
||||
AC122371000
|
Thermo Scientific Chemicals
122371000 |
100 g | Glass Bottle |
Each for $109.90
|
|
||||
AC122370025
|
Thermo Scientific Chemicals
122370025 |
2.5 kg | Glass bottle |
Each for $1,443.96
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Alkylation reagentChemical Identifiers
74-88-4 | |
141.94 | |
INQOMBQAUSQDDS-UHFFFAOYSA-N | |
6328 | |
iodomethane |
CH3I | |
MFCD00001073 | |
methyl iodide, monoiodomethane, methane, iodo, methyljodid, jod-methan, methyliodide, methyljodide, iodometano, joodmethaan, metylu jodek | |
CHEBI:39282 | |
CI |
Specifications
74-88-4,7440-50-8 | |
2.2800g/mL | |
Authentic | |
Glass bottle | |
1.5290 to 1.5310 | |
MFCD00001073 | |
01, 69 | |
2.28 | |
methyl iodide, monoiodomethane, methane, iodo, methyljodid, jod-methan, methyliodide, methyljodide, iodometano, joodmethaan, metylu jodek | |
INQOMBQAUSQDDS-UHFFFAOYSA-N | |
iodomethane | |
Copper | |
0.5 mPa.s (20°C) | |
141.94 | |
Liquid |
-66.0°C | |
42.5°C | |
98.5% min. (GC) | |
CH3I | |
CH3I | |
5 g | |
01,682; 02,274; 03,202; 04,341; 05,447; 06,384; 08,342; 09,308; 15,118 | |
15, 6159 | |
Solubility in water: 14g/L (20°C). Other solubilities: miscible with alcohol and ether, soluble in benzene, acetone, methylenechloride, n,n-dimethylformamide | |
CI | |
141.94 | |
6328 | |
CHEBI:39282 | |
99% | |
Iodomethane |
Safety and Handling
Emergency Overview
Limited evidence of a carcinogenic effect. Toxic by inhalation and if swallowed. Harmful in contact with skin. Irritating to eyes, respiratory system and skin.
NFPA
Signal Word: WARNING!
RUO – Research Use Only