missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Iohexol
CAS: 66108-95-0 | C19H26I3N3O9, C19H26I3N3O9 | 821.14 g/mol
$774.02 - $774.02
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC466651000
|
Thermo Scientific Chemicals
466651000 |
100 g |
Each for $774.02
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 66108-95-0 | |
| C19H26I3N3O9 |
Specifications
| 66108-95-0 | |
| Solubility aq. soln.: soluble. | |
| C19H26I3N3O9 | |
| NTHXOOBQLCIOLC-UHFFFAOYNA-N | |
| N1,N3-bis(2,3-dihydroxypropyl)-5-[N-(2,3-dihydroxypropyl)acetamido]-2,4,6-triiodobenzene-1,3-dicarboxamide | |
| Iohexol |
| 100 g | |
| 180.0°C | |
| 821.13 | |
| CC(=O)N(CC(O)CO)C1=C(I)C(C(=O)NCC(O)CO)=C(I)C(C(=O)NCC(O)CO)=C1I | |
| 821.14 |
Safety and Handling
EINECSNumber : 266-164-2
RUO – Research Use Only