missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ionic Strength Adjustment Sodium Hydroxide Buffer, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Supplier: SPEX CertiPrep Group LLC ISBUF2500
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
| Analyte | Concentration |
|---|---|
| NaOH | 10M |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
| 10M Sodium Hydroxide Buffer Solution | |
| 10 M | |
| Liquid | |
| Standard |
| 1310-73-2,7732-18-5 | |
| pH Calibration | |
| 500 mL | |
| Triple-leached LDPE bottle |
Product intended for laboratory use only