missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sartorius Ionizer with Power Supply

Ionizer with 115 V Power Supply.

Supplier:  Sartorius YIB02115V

Catalog No. 14-558-727


Explore available promotions

Description

Description

  • Ionizer with U-Shaped Electrode
  • Includes 115V power supply
  • For use with Cubis™ II
  • Made by HAUG
Specifications

Specifications

Cubis II
115 V Power Supply
Power Supply
Videos
SDS
Documents

Documents

Promotions

Promotions