missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ionomycin, calcium salt, 100%, >98%, For HPLC analysis, MP Biomedicals™
$238.79 - $238.79
Chemical Identifiers
CAS | 56092-82-1 |
---|---|
Molecular Formula | C41H70CaO9 |
Molecular Weight (g/mol) | 747.08 |
MDL Number | MFCD00083634 |
InChI Key | RULWQOBWZOQCOY-SQNLDGQHSA-M |
PubChem CID | 71311740 |
IUPAC Name | calcium;(4R,6S,8S,10E,12R,14R,16E,18R,19R,20S,21S)-19,21-dihydroxy-22-[(2S,5S)-5-[(2R,5S)-5-[(1R)-1-hydroxyethyl]-5-methyloxolan-2-yl]-5-methyloxolan-2-yl]-4,6,8,12,14,18,20-heptamethyl-11-oxido-9-oxodocosa-10,16-dienoate |
SMILES | [Ca++].C[C@H](CCC([O-])=O)C[C@H](C)C[C@H](C)C(=O)[CH-]C(=O)[C@H](C)C[C@H](C)C\C=C\[C@@H](C)[C@@H](O)[C@@H](C)[C@@H](O)C[C@@H]1CC[C@](C)(O1)[C@H]1CC[C@](C)(O1)[C@@H](C)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
ICN15507001
|
MP Biomedicals Inc
0215507001 |
1 mg |
Each for $238.79
|
|
|||||
Chemical Identifiers
56092-82-1 | |
747.08 | |
RULWQOBWZOQCOY-SQNLDGQHSA-M | |
calcium;(4R,6S,8S,10E,12R,14R,16E,18R,19R,20S,21S)-19,21-dihydroxy-22-[(2S,5S)-5-[(2R,5S)-5-[(1R)-1-hydroxyethyl]-5-methyloxolan-2-yl]-5-methyloxolan-2-yl]-4,6,8,12,14,18,20-heptamethyl-11-oxido-9-oxodocosa-10,16-dienoate |
C41H70CaO9 | |
MFCD00083634 | |
71311740 | |
[Ca++].C[C@H](CCC([O-])=O)C[C@H](C)C[C@H](C)C(=O)[CH-]C(=O)[C@H](C)C[C@H](C)C\C=C\[C@@H](C)[C@@H](O)[C@@H](C)[C@@H](O)C[C@@H]1CC[C@](C)(O1)[C@H]1CC[C@](C)(O1)[C@@H](C)O |
Specifications
Ionomycin Calcium Salt | |
56092-82-1 | |
1 mg | |
C41H70CaO9 | |
RULWQOBWZOQCOY-SQNLDGQHSA-M | |
calcium;(4R,6S,8S,10E,12R,14R,16E,18R,19R,20S,21S)-19,21-dihydroxy-22-[(2S,5S)-5-[(2R,5S)-5-[(1R)-1-hydroxyethyl]-5-methyloxolan-2-yl]-5-methyloxolan-2-yl]-4,6,8,12,14,18,20-heptamethyl-11-oxido-9-oxodocosa-10,16-dienoate | |
71311740 | |
>98% |
Crystalline Powder | |
White | |
1 | |
MFCD00083634 | |
[Ca++].C[C@H](CCC([O-])=O)C[C@H](C)C[C@H](C)C(=O)[CH-]C(=O)[C@H](C)C[C@H](C)C\C=C\[C@@H](C)[C@@H](O)[C@@H](C)[C@@H](O)C[C@@H]1CC[C@](C)(O1)[C@H]1CC[C@](C)(O1)[C@@H](C)O | |
747.08 | |
747.1 | |
Ionomycin, calcium salt, For HPLC analysis |