Tingley™ Iron Eagle™ Polyurethane Bib Overalls
Extremely comfortable and flexible. Resists fats, oils, pine tars, gasoline, grease and a variety of other chemicals.
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
29-635-2558
|
Tingley™
O22007XL |
X-Large |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
- Lightweight and durable
- Resists fats, oils, pine tars, gasoline, grease and a variety of other chemicals
- Snag and abrasion resistant
- Heat sealed seams
- Therm-O-Rad™ sealed seams
- Storm fly front
- Premium snap-lock suspender buckles
- Heavy-duty suspenders with crossover
- Heavy-duty snaps
- Mildew resistant
Order Info
Contact your Fisher Safety Customer Service Representative for ordering information on sizes not shown.
Specifications
Snap-lock | |
Bib Overalls |