missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Iron Filings

Iron Filings are great for magnetism demonstrations.

$25.83 - $32.20

Specifications

Description Iron Filings
Material 100% Iron
Shape Cylindrical
For Use With (Application) For Magnetism Demonstrations
Grade 9 to 12
View More Specs

Products 3
Catalog Number Mfr. No. Product Type Price Quantity  
Catalog Number Mfr. No. Product Type Price Quantity  
S95969
View Documents
Eisco™
PH0799B
Fine Iron Filing, Sprinkle Jar
Each for $25.83
Only null left
 
S95968
View Documents
Eisco™
PH0799A
Coarse Iron Filing
Each for $27.47
Only null left
 
S95970
View Documents
Eisco™
PH0799C
Fine Iron Filing, Plastic Jar
Each for $32.20
Only null left
 
Description

Description

  • Choose from coarse or fine grain size
  • Convenient packaging choices
Specifications

Specifications

Iron Filings
Cylindrical
9 to 12
GHS-compliant label printed with safety and storage information
100% Iron
For Magnetism Demonstrations
Grey
8 years old
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'