missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Iron(II) acetate, 95%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 305370500

 View more versions of this product

Catalog No. AC305370500


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Iron(II) acetate
95%
Brown or Gray to White
50 g
C4H6FeO4
MFCD00058909
LNOZJRCUHSPCDZ-UHFFFAOYSA-L
iron(2+);diacetate
18344
95%
Glass bottle
190.0°C
3094-87-9
Powder
95%
(CH3CO2)2
iron ii acetate, ferrous acetate, iron diacetate, iron di acetate, iron 2+ acetate, acetic acid, iron 2+ salt, acetic acid, iron 2+ salt 2:1, unii-l80i7m6d3q, ferrousacetate
[Fe++].CC([O-])=O.CC([O-])=O
173.93
173.93
Authentic

Chemical Identifiers

3094-87-9
173.93
LNOZJRCUHSPCDZ-UHFFFAOYSA-L
18344
C4H6FeO4
MFCD00058909
iron ii acetate, ferrous acetate, iron diacetate, iron di acetate, iron 2+ acetate, acetic acid, iron 2+ salt, acetic acid, iron 2+ salt 2:1, unii-l80i7m6d3q, ferrousacetate
[Fe++].CC([O-])=O.CC([O-])=O
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 221-441-7

RTECSNumber : AI3850000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only