missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Iron(II) chloride tetrahydrate, 98%, Thermo Scientific Chemicals

$123.10 - $370.71

Chemical Identifiers

CAS 13478-10-9
Molecular Formula Cl2FeH8O4
Molecular Weight (g/mol) 198.805
MDL Number MFCD00149709
InChI Key WSSMOXHYUFMBLS-UHFFFAOYSA-L
Synonym ferrous chloride tetrahydrate, iron ii chloride tetrahydrate, iron dichloride tetrahydrate, cl2fe.4h2o, fecl2.4h2o, iron chloride, tetrahydrate, dichloroiron-water 1/4, iron ii chloride tetra hydrate, ae>> nciu eae r masculineiii
PubChem CID 16211588
ChEBI CHEBI:86249
IUPAC Name dichloroiron;tetrahydrate
SMILES O.O.O.O.Cl[Fe]Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1632730
View Documents
Thermo Scientific Chemicals
A1632730
250 g
Each for $123.10
 
AAA163270B
View Documents
Thermo Scientific Chemicals
A163270B
1000 g
Each for $370.71
 
Description

Description

Iron(II) chloride tetrahydrate is used as a reducing agent in metallurgy, in pharmaceutical preparations, as a mordant in dyeing and in sewage treatment. Ferrous chloride has a kind of niche application. During the laboratory synthesis of iron complexes, ferrous chloride acts as a reducing flocculating agent in waste water treatment, particularly for wastes containing chromate. It is the precursor to hydrated iron(III) oxides that are magnetic pigments. Ferrous chloride is employed as a reducing agent in many organic synthesis reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Iron(II) chloride tetrahydrate is used as a reducing agent in metallurgy, in pharmaceutical preparations, as a mordant in dyeing and in sewage treatment. Ferrous chloride has a kind of niche application. During the laboratory synthesis of iron complexes, ferrous chloride acts as a reducing flocculating agent in waste water treatment, particularly for wastes containing chromate. It is the precursor to hydrated iron(III) oxides that are magnetic pigments. Ferrous chloride is employed as a reducing agent in many organic synthesis reactions.

Solubility
Soluble in water.

Notes
Air and light-sensitive. Hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong base, strong acids, ethylene oxide and strong oxidizing agents.
Specifications

Chemical Identifiers

13478-10-9
198.805
WSSMOXHYUFMBLS-UHFFFAOYSA-L
16211588
dichloroiron;tetrahydrate
Cl2FeH8O4
MFCD00149709
ferrous chloride tetrahydrate, iron ii chloride tetrahydrate, iron dichloride tetrahydrate, cl2fe.4h2o, fecl2.4h2o, iron chloride, tetrahydrate, dichloroiron-water 1/4, iron ii chloride tetra hydrate, ae>> nciu eae r masculineiii
CHEBI:86249
O.O.O.O.Cl[Fe]Cl

Specifications

13478-10-9
Cl2FeH8O4
MFCD00149709
14,4043
Soluble in water.
O.O.O.O.Cl[Fe]Cl
198.805
CHEBI:86249
98%
Air sensitive; Hygroscopic
Iron(II) chloride tetrahydrate
250 g
FeCl2·4H2O
UN3260
ferrous chloride tetrahydrate, iron ii chloride tetrahydrate, iron dichloride tetrahydrate, cl2fe.4h2o, fecl2.4h2o, iron chloride, tetrahydrate, dichloroiron-water 1/4, iron ii chloride tetra hydrate, ae>> nciu eae r masculineiii
WSSMOXHYUFMBLS-UHFFFAOYSA-L
dichloroiron;tetrahydrate
16211588
198.81 (126.75 Anhydrous)
Odorless
1.93
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.

P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302-H314-H335

DOTInformation : Hazard Class: 8; Packaging Group: II

EINECSNumber : 231-843-4

RTECSNumber : NO5600000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only