Learn More
Iron(II) fluoride, anhydrous, 98%, Thermo Scientific Chemicals
$139.81 - $470.33
Chemical Identifiers
CAS | 7789-28-8 |
---|---|
Molecular Formula | F2Fe |
Molecular Weight (g/mol) | 93.84 |
MDL Number | MFCD00016093 |
InChI Key | FZGIHSNZYGFUGM-UHFFFAOYSA-L |
IUPAC Name | λ²-iron(2+) difluoride |
SMILES | [F-].[F-].[Fe++] |
Description
Iron(II) fluoride is used for proteomics research applications. It is also a chemical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7789-28-8 | |
93.84 | |
FZGIHSNZYGFUGM-UHFFFAOYSA-L | |
[F-].[F-].[Fe++] |
F2Fe | |
MFCD00016093 | |
λ²-iron(2+) difluoride |
Specifications
>1000°C (sublimation) | |
White | |
10 g | |
MFCD00016093 | |
14,4045 | |
FZGIHSNZYGFUGM-UHFFFAOYSA-L | |
λ²-iron(2+) difluoride | |
93.84 | |
Air sensitive; Hygroscopic | |
Iron(II) fluoride, Anhydrous |
7789-28-8 | |
Powder | |
F2Fe | |
UN3260 | |
Insoluble in water. | |
[F-].[F-].[Fe++] | |
93.84 | |
98% | |
4.09 g/mL |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
EINECSNumber : 232-155-7
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only