Learn More
Iron(II) oxalate dihydrate, 99%, Thermo Scientific Chemicals
$84.53 - $284.68
Chemical Identifiers
CAS | 6047-25-2 |
---|---|
Molecular Formula | C2H4FeO6 |
Molecular Weight (g/mol) | 179.89 |
MDL Number | MFCD00150040 MFCD00012475 |
InChI Key | NPLZZSLZTJVZSX-UHFFFAOYSA-L |
Synonym | ferrous oxalate dihydrate, unii-z6x3ybu50d, z6x3ybu50d, iron ii oxalate hydrate, iron ii oxalate dihydrate, iron oxalate dihydrate, iron 2+ ; oxalate; dihydrate, iron, diaqua ethanedioato 2--kappao1,kappao2-, t-4, iron ii oxalate dihydrate 250g, iron ii oxalate dihydrate, puratronic™ |
PubChem CID | 516788 |
IUPAC Name | iron(2+);oxalate;dihydrate |
SMILES | O.O.[Fe++].[O-]C(=O)C([O-])=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1347930
|
Thermo Scientific Chemicals
A1347930 |
250 g |
Each for $84.53
|
|
|||||
AAA134790B
|
Thermo Scientific Chemicals
A134790B |
1000 g |
Each for $284.68
|
|
|||||
Description
It is used as photographic developer for silver bromide-gelatin plates. It imparts greenish-brown tint to optical glass (sunglasses, windshields, railroad car windows), for decorative glassware, pigment for plastics, paints and lacquers. It acts as reagent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used as photographic developer for silver bromide-gelatin plates. It imparts greenish-brown tint to optical glass (sunglasses, windshields, railroad car windows), for decorative glassware, pigment for plastics, paints and lacquers. It acts as reagent.
Solubility
Soluble in water. Insoluble in acetic acid.
Notes
Store at room temperature. It is not intended for diagnostic and therapeutic purposes. Avoid contamination with oxidizing agents.
Chemical Identifiers
6047-25-2 | |
179.89 | |
NPLZZSLZTJVZSX-UHFFFAOYSA-L | |
516788 | |
O.O.[Fe++].[O-]C(=O)C([O-])=O |
C2H4FeO6 | |
MFCD00150040 MFCD00012475 | |
ferrous oxalate dihydrate, unii-z6x3ybu50d, z6x3ybu50d, iron ii oxalate hydrate, iron ii oxalate dihydrate, iron oxalate dihydrate, iron 2+ ; oxalate; dihydrate, iron, diaqua ethanedioato 2--kappao1,kappao2-, t-4, iron ii oxalate dihydrate 250g, iron ii oxalate dihydrate, puratronic™ | |
iron(2+);oxalate;dihydrate |
Specifications
190°C (decomposition) | |
250 g | |
C2H4FeO6 | |
ferrous oxalate dihydrate, unii-z6x3ybu50d, z6x3ybu50d, iron ii oxalate hydrate, iron ii oxalate dihydrate, iron oxalate dihydrate, iron 2+ ; oxalate; dihydrate, iron, diaqua ethanedioato 2--kappao1,kappao2-, t-4, iron ii oxalate dihydrate 250g, iron ii oxalate dihydrate, puratronic™ | |
NPLZZSLZTJVZSX-UHFFFAOYSA-L | |
iron(2+);oxalate;dihydrate | |
516788 | |
99% | |
2.28 |
6047-25-2 | |
99% | |
MFCD00150040 MFCD00012475 | |
Soluble in water. Insoluble in acetic acid. | |
O.O.[Fe++].[O-]C(=O)C([O-])=O | |
179.89 | |
179.90 (143.87 Anhydrous) | |
Hygroscopic | |
Iron (II) Oxalate Dihydrate |
Safety and Handling
GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.
P264b-P270-P280-P301+P312-P302+P352-P312-P330-P363-P501c
H302+H312
EINECSNumber : 208-217-4
TSCA : Yes
storageNote1 : Hygroscopic
Recommended Storage : Ambient temperatures
RUO – Research Use Only