missing translation for 'onlineSavingsMsg'
Learn More

Iron(II) oxide, 99.5% (metals basis)

In manufacture of green, heat-absorbing glass | CAS: 1345-25-1 | FeO | 71.844 g/mol

$310.70 - $1,244.53

Chemical Identifiers

CAS 1345-25-1
Molecular Formula FeO
Molecular Weight (g/mol) 71.844
MDL Number MFCD00016095
InChI Key UQSXHKLRYXJYBZ-UHFFFAOYSA-N
Synonym iron ii oxide, ferrous oxide, iron monooxide, iron monoxide, iron oxide, wustite, natural wuestite, eisenoxydul, hsdb 464, eisen ii-oxid
PubChem CID 14945
ChEBI CHEBI:50820
IUPAC Name oxoiron
SMILES O=[Fe]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA3051306
View Documents
Thermo Scientific Chemicals
03051306
5 g N/A
Only null left
 
AA3051314
View Documents
Thermo Scientific Chemicals
03051314
25 g N/A
Only null left
 
Description

Description

In manufacture of green, heat-absorbing glass; in steel manufacture; and as a catalystIron(II) oxide is used in manufacturing of green, heat-absorbing glass; in steel manufacture; and as a catalyst. It is used as a dye or pigment in pottery, glazes and glasses. It finds application as heat-absorbing glass used in buildings and automobiles. It is used as a precursor in the production of steel. It is also used in cosmetics and some tattoo inks. It can also be used for filtering phosphates from home aquaria and acts as a catalyst in a number of industrial and chemical operations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1345-25-1
71.844
UQSXHKLRYXJYBZ-UHFFFAOYSA-N
14945
oxoiron
FeO
MFCD00016095
iron ii oxide, ferrous oxide, iron monooxide, iron monoxide, iron oxide, wustite, natural wuestite, eisenoxydul, hsdb 464, eisen ii-oxid
CHEBI:50820
O=[Fe]

Specifications

1369°C
5 g
FeO
iron ii oxide, ferrous oxide, iron monooxide, iron monoxide, iron oxide, wustite, natural wuestite, eisenoxydul, hsdb 464, eisen ii-oxid
UQSXHKLRYXJYBZ-UHFFFAOYSA-N
oxoiron
14945
71.85
5.7
1345-25-1
99.5% (metals basis)
MFCD00016095
Insoluble in water and alkalies. Soluble in HCl,H2SO4. Slightly soluble in HNO3Insoluble in water,alcohol and alkali. Soluble in hydrochloric acid and sulfuric acid. Slightly soluble in nitric acid.
O=[Fe]
71.844
CHEBI:50820
Air Sensitive
Iron(II) oxide
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 215-721-8

TSCA : Yes

storageNote1 : Air Sensitive

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only