missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Iron(II) sulfate heptahydrate, 99.5%, for analysis, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 201390051

 View more versions of this product

Catalog No. AC201390051


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Iron(II) sulfate heptahydrate
7782-63-0
Crystals
99.5%
FeSO4·7H2O
01,393; 04,237; 05,308
iron ii sulfate heptahydrate, ferrous sulfate heptahydrate, presfersul, melanterite mineral, iron sulfate heptahydrate, iron 2+ sulfate heptahydrate, fesofor, fesotyme, haemofort, ironate
SURQXAFEQWPFPV-UHFFFAOYSA-L
iron(2+);sulfate;heptahydrate
62662
278.01
Analytical
64.0°C
Blue to Green
5 kg
FeH14O11S
MFCD00149719
15,4084
Solubility in water: 25.6 g/100mL (20°C). Other solubilities: insoluble in alcohol
O.O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O
278.01
CHEBI:75836
99.5%
Plastic bottle

Chemical Identifiers

7782-63-0
278.01
SURQXAFEQWPFPV-UHFFFAOYSA-L
62662
O.O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O
FeH14O11S
MFCD00149719
iron ii sulfate heptahydrate, ferrous sulfate heptahydrate, presfersul, melanterite mineral, iron sulfate heptahydrate, iron 2+ sulfate heptahydrate, fesofor, fesotyme, haemofort, ironate
CHEBI:75836
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.

GHS P Statement
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only