missing translation for 'onlineSavingsMsg'
Learn More

Iron(III) bromide, anhydrous, 98+%

CAS: 10031-26-2

Supplier:  Thermo Scientific Chemicals 03112522

 View more versions of this product

Catalog No. AA3112522


Only null left
Explore available promotions

Description

Description

Iron(III) bromide is used as a Lewis acid catalyst in the halogenation of aromatic compounds. It is also used in the preparation of 2,5-dibromo-4-aryl-1,3-pentadiene by reacting with homoallenic bromohydrin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Iron(III) bromide
Anhydrous
10031-26-2
100 g
14,4018
295.57
Odorless
(decomposition)
Red
Crystalline
MFCD00016082
Soluble in water,ether and alcohol.
≥98%
Hygroscopic; Light sensitive

Chemical Identifiers

10031-26-2
MFCD00016082
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H302-H335-H319
Causes severe skin burns and eye damage.
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 233-089-1

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only