missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Iron(III) chloride, 98%, pure, anhydrous, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 169430010

 View more versions of this product

Catalog No. AC169430010


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid
Specifications

Specifications

Iron(III) chloride
Gray to Black
1 kg
Cl3Fe
MFCD00011005
15,4046
Solubility in water: 480g/L (20°C). Other solubilities: readily soluble in alcohol, ether and acetone, slightly soluble in cs2, practically insoluble in ethyl acetate
[Cl-].[Cl-].[Cl-].[Fe+3]
162.20
98%
Plastic bottle
7705-08-0
Powder
97.5 to 102.5% (Iodometry)
FeCl3
01,392; 02,199; 04,236; 05,307; 06,259; 07,153; 08,228; 09,222; 10,185; 11,237; 12,1; 13,133; 14,164; 16,167; 17,138
Ferric chloride
RBTARNINKXHZNM-UHFFFAOYSA-K
iron(3+) trichloride
162.21
Pure

Chemical Identifiers

7705-08-0
162.20
RBTARNINKXHZNM-UHFFFAOYSA-K
iron(3+) trichloride
Cl3Fe
MFCD00011005
Ferric chloride
[Cl-].[Cl-].[Cl-].[Fe+3]
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes serious eye damage.
Causes skin irritation.
May cause an allergic skin reaction.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present

GHS Signal Word: Danger

EINECSNumber : 231-729-4

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only