Learn More
Iron(III) chloride hexahydrate, 97%, Thermo Scientific Chemicals
$114.68 - $1572.34
Chemical Identifiers
CAS | 10025-77-1 |
---|---|
Molecular Formula | Cl3FeH12O6 |
Molecular Weight (g/mol) | 270.29 |
MDL Number | MFCD00149712 |
InChI Key | NQXWGWZJXJUMQB-UHFFFAOYSA-K |
Synonym | Ferric chloride hexahydrate |
IUPAC Name | iron(3+) hexahydrate trichloride |
SMILES | O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Fe+3] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAA1623136
|
Thermo Scientific Chemicals
A1623136 |
500 g |
Each for $114.68
|
|
||||||
AAA162310E
|
Thermo Scientific Chemicals
A162310E |
2500 g |
Each for $456.55
|
|
||||||
AAA162310C
|
Thermo Scientific Chemicals
A162310C |
10,000 g |
Each for $1,572.34
|
|
||||||
Description
Iron(III) chloride hexahydrate is used to prepare organically templated iron phosphates. It is used for sewerage treatment of industrial wastes, chloride hydrometallurgy (Silgarin process for the production of silicon), drinking water production, production of precursors for polyvinyl chloride (PVC) and copper-based metals in printed circuit boards. It plays an important role in photoengraving and photography as an etching agent. It is useful as a mordant in dyeing and printing textiles. It is useful in the detection of phenols and phenolic derivatives; gamma-hydroxybutyric acids and also in the Trinder spot test for detecting salicylic acids.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIron(III) chloride hexahydrate is used to prepare organically templated iron phosphates. It is used for sewerage treatment of industrial wastes, chloride hydrometallurgy (Silgarin process for the production of silicon), drinking water production, production of precursors for polyvinyl chloride (PVC) and copper-based metals in printed circuit boards. It plays an important role in photoengraving and photography as an etching agent. It is useful as a mordant in dyeing and printing textiles. It is useful in the detection of phenols and phenolic derivatives; gamma-hydroxybutyric acids and also in the Trinder spot test for detecting salicylic acids.
Solubility
Soluble in water, alcohol, glycerol, methanol, ether and acetone. Insoluble in ethyl acetate.
Notes
Hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, sodium and sodium oxides and potassium.
Chemical Identifiers
10025-77-1 | |
270.29 | |
NQXWGWZJXJUMQB-UHFFFAOYSA-K | |
iron(3+) hexahydrate trichloride |
Cl3FeH12O6 | |
MFCD00149712 | |
Ferric chloride hexahydrate | |
O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Fe+3] |
Specifications
280°C to 285°C | |
10025-77-1 | |
500 g | |
FeCl3·6H2O | |
UN3260 | |
Ferric chloride hexahydrate | |
NQXWGWZJXJUMQB-UHFFFAOYSA-K | |
iron(3+) hexahydrate trichloride | |
270.30 (162.21 Anhydrous) | |
Odorless | |
1.82 |
37°C | |
1.8 | |
Cl3FeH12O6 | |
MFCD00149712 | |
14,4019 | |
Soluble in water,alcohol,glycerol,methanol,ether and acetone. Insoluble in ethyl acetate. | |
O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Fe+3] | |
270.29 | |
97% | |
Hygroscopic | |
Iron(III) chloride hexahydrate |
Safety and Handling
GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
P260-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P314-P330-P333+P313-P362-P501c
H302-H315-H317-H318-H373
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
EINECSNumber : 231-729-4
RTECSNumber : NO5425000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only