Learn More
Iron(III) fluoride, anhydrous, 97% min, Thermo Scientific Chemicals
$77.44 - $262.97
Chemical Identifiers
CAS | 7783-50-8 |
---|---|
Molecular Formula | F3Fe |
Molecular Weight (g/mol) | 112.84 |
MDL Number | MFCD00011012 |
InChI Key | SHXXPRJOPFJRHA-UHFFFAOYSA-K |
Synonym | ferric fluoride, iron iii fluoride, iron fluoride, iron trifluoride, iron fluoride fef3, ferric trifluoride, fef3, hsdb 450, iron iii fluoride, anhydrous, ferric fluoride, anhydrous |
PubChem CID | 24552 |
IUPAC Name | trifluoroiron |
SMILES | F[Fe](F)F |
Description
Iron(III) fluroide is used in ceramics manufacturing. It is used as a catalyst for certain cross coupling reactions. Further, it is involved in the chemoselective addition of cyanide to aldehyde to prepare cyanohydrins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7783-50-8 | |
112.84 | |
SHXXPRJOPFJRHA-UHFFFAOYSA-K | |
24552 | |
F[Fe](F)F |
F3Fe | |
MFCD00011012 | |
ferric fluoride, iron iii fluoride, iron fluoride, iron trifluoride, iron fluoride fef3, ferric trifluoride, fef3, hsdb 450, iron iii fluoride, anhydrous, ferric fluoride, anhydrous | |
trifluoroiron |
Specifications
>1000° (sublimation) | |
7783-50-8 | |
10 g | |
F3Fe | |
ferric fluoride, iron iii fluoride, iron fluoride, iron trifluoride, iron fluoride fef3, ferric trifluoride, fef3, hsdb 450, iron iii fluoride, anhydrous, ferric fluoride, anhydrous | |
SHXXPRJOPFJRHA-UHFFFAOYSA-K | |
trifluoroiron | |
24552 | |
Hygroscopic | |
3.87 g/cm3 |
Green | |
Powder | |
97% min. (Metal basis) | |
MFCD00011012 | |
Soluble in dilute hydrofluroic acid. Slightly soluble in water. Insoluble in alcohol,ether and benzene. | |
F[Fe](F)F | |
112.84 | |
112.84 | |
Pale green powder | |
Iron(III) fluoride |
Safety and Handling
H301-H311-H332-H314-H318: Toxic if swallowed. Toxic in contact with skin. Harmful if inhaled. Causes severe skin burns and eye damage. Causes serious eye damage.
EINECSNumber : 232-002-4
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only