missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Iron(III) hydroxide, 99+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1626730

 View more versions of this product

Catalog No. AAA1626730


Explore available promotions

Description

Description

Iron(III) hydroxide, alpha-phase, is used as a catalyst for proteomics research. It is also used as a pharamaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Iron(III) hydroxide, alpha-phase, is used as a catalyst for proteomics research. It is also used as a pharamaceutical intermediate.

Solubility
It is insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
Specifications

Specifications

Iron(III) hydroxide
20344-49-4
FeHO2
MFCD00064782
hydroxy oxo iron, goethite, lepidocrocite, iron hydroxide oxide, goethite fe oh o, iron hydroxide oxide fe oh o, iron iii oxide-hydroxide, ferric acid, iron pigment yellow, ferric hydroxide oxide
AEIXRCIKZIZYPM-UHFFFAOYSA-M
hydroxy(oxo)iron
91502
≥99%
4.1
alpha-phase
250 g
FeO(OH)
14,4025
It is insoluble in water.
O[Fe]=O
88.851
88.85
Odorless

Chemical Identifiers

20344-49-4
88.851
AEIXRCIKZIZYPM-UHFFFAOYSA-M
91502
O[Fe]=O
FeHO2
MFCD00064782
hydroxy oxo iron, goethite, lepidocrocite, iron hydroxide oxide, goethite fe oh o, iron hydroxide oxide fe oh o, iron iii oxide-hydroxide, ferric acid, iron pigment yellow, ferric hydroxide oxide
hydroxy(oxo)iron
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 243-746-4

RTECSNumber : NO7400000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only