missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Iron(III) meso-tetraphenylporphine chloride, Thermo Scientific Chemicals
$94.67 - $310.25
Chemical Identifiers
CAS | 16456-81-8 |
---|---|
Molecular Formula | C44H30Cl3FeN4 |
Molecular Weight (g/mol) | 776.947 |
MDL Number | MFCD00012152 |
InChI Key | QDTNPIPZGCOFPA-UHFFFAOYSA-K |
Synonym | 5,10,15,20-tetraphenyl-21h,23h-porphine iron iii chloride, acmc-20am7m, chloro 5,10,15,20-tetraphenyl porphyrinato iron iii, iron iii meso-tetraphenylporphine chloride |
PubChem CID | 67043998 |
IUPAC Name | iron(3+);5,10,15,20-tetraphenyl-21,22-dihydroporphyrin;trichloride |
SMILES | C1=CC=C(C=C1)C2=C3C=CC(=C(C4=CC=C(N4)C(=C5C=CC(=N5)C(=C6C=CC2=N6)C7=CC=CC=C7)C8=CC=CC=C8)C9=CC=CC=C9)N3.[Cl-].[Cl-].[Cl-].[Fe+3] |
Description
Iron(III) meso-tetraphenylporphine chloride is used catalyzing reagent for silylation reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
16456-81-8 | |
776.947 | |
QDTNPIPZGCOFPA-UHFFFAOYSA-K | |
67043998 | |
C1=CC=C(C=C1)C2=C3C=CC(=C(C4=CC=C(N4)C(=C5C=CC(=N5)C(=C6C=CC2=N6)C7=CC=CC=C7)C8=CC=CC=C8)C9=CC=CC=C9)N3.[Cl-].[Cl-].[Cl-].[Fe+3] |
C44H30Cl3FeN4 | |
MFCD00012152 | |
5,10,15,20-tetraphenyl-21h,23h-porphine iron iii chloride, acmc-20am7m, chloro 5,10,15,20-tetraphenyl porphyrinato iron iii, iron iii meso-tetraphenylporphine chloride | |
iron(3+);5,10,15,20-tetraphenyl-21,22-dihydroporphyrin;trichloride |
Specifications
16456-81-8 | |
250 mg | |
MFCD00012152 | |
Insoluble in water. | |
C1=CC=C(C=C1)C2=C3C=CC(=C(C4=CC=C(N4)C(=C5C=CC(=N5)C(=C6C=CC2=N6)C7=CC=CC=C7)C8=CC=CC=C8)C9=CC=CC=C9)N3.[Cl-].[Cl-].[Cl-].[Fe+3] | |
776.947 | |
704.04 |
Crystalline | |
C44H30Cl3FeN4 | |
5,10,15,20-tetraphenyl-21h,23h-porphine iron iii chloride, acmc-20am7m, chloro 5,10,15,20-tetraphenyl porphyrinato iron iii, iron iii meso-tetraphenylporphine chloride | |
QDTNPIPZGCOFPA-UHFFFAOYSA-K | |
iron(3+);5,10,15,20-tetraphenyl-21,22-dihydroporphyrin;trichloride | |
67043998 | |
Iron(III) meso-tetraphenylporphine chloride |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only