missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Iron(III) Stearate, TCI America™
$96.22 - $96.22
Chemical Identifiers
CAS | 555-36-2 |
---|---|
Molecular Formula | C54H105FeO6 |
Molecular Weight (g/mol) | 906.273 |
MDL Number | MFCD00050716 |
InChI Key | XHQSLVIGPHXVAK-UHFFFAOYSA-K |
Synonym | Ferric Stearate, Stearic Acid Iron(III) Salt |
PubChem CID | 68388 |
IUPAC Name | iron(3+);octadecanoate |
SMILES | CCCCCCCCCCCCCCCCCC(=O)[O-].CCCCCCCCCCCCCCCCCC(=O)[O-].CCCCCCCCCCCCCCCCCC(=O)[O-].[Fe+3] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
S025625G
|
TCI America
S025625G |
25 g |
Each for $96.22
|
|
|||||
Chemical Identifiers
555-36-2 | |
906.273 | |
XHQSLVIGPHXVAK-UHFFFAOYSA-K | |
68388 | |
CCCCCCCCCCCCCCCCCC(=O)[O-].CCCCCCCCCCCCCCCCCC(=O)[O-].CCCCCCCCCCCCCCCCCC(=O)[O-].[Fe+3] |
C54H105FeO6 | |
MFCD00050716 | |
Ferric Stearate, Stearic Acid Iron(III) Salt | |
iron(3+);octadecanoate |
Specifications
555-36-2 | |
MFCD00050716 | |
Ferric Stearate, Stearic Acid Iron(III) Salt | |
CCCCCCCCCCCCCCCCCC(=O)[O-].CCCCCCCCCCCCCCCCCC(=O)[O-].CCCCCCCCCCCCCCCCCC(=O)[O-].[Fe+3] | |
906.273 | |
906.27 | |
Iron(III) Stearate |
C54H105FeO6 | |
25 g | |
XHQSLVIGPHXVAK-UHFFFAOYSA-K | |
iron(3+);octadecanoate | |
68388 | |
Crystalline Powder |
Safety and Handling
EINECSNumber : (2)-0625
TSCA : Yes