Learn More
Iron(III) sulfate hydrate, Reagent Grade, Thermo Scientific Chemicals
$91.44 - $600.00
Chemical Identifiers
CAS | 15244-10-7 |
---|---|
Molecular Formula | Fe2H2O13S3 |
Molecular Weight (g/mol) | 417.87 |
MDL Number | MFCD00011007 |
InChI Key | VXWSFRMTBJZULV-UHFFFAOYSA-H |
PubChem CID | 167265 |
SMILES | O.[Fe+3].[Fe+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA3331630
|
Thermo Scientific Chemicals
03331630 |
250 g |
Each for $91.44
|
|
|||||
AA33316A1
|
Thermo Scientific Chemicals
033316A1 |
1 kg |
Each for $230.10
|
|
|||||
AA33316A7
|
Thermo Scientific Chemicals
033316A7 |
5 kg |
Each for $600.00
|
|
|||||
Description
Iron(III) sulfate is also used in pigments, pickling baths for aluminum and steel. It acts as a mordant in dyeing, as a coagulant for industrial wastes. In the medical field, it is used as an astringent and styptic. It is involved in the preparation of aromatic esters from the corresponding acids and alcohols. Further, it is employed in the oxidation of pyrite.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
15244-10-7 | |
417.87 | |
VXWSFRMTBJZULV-UHFFFAOYSA-H | |
O.[Fe+3].[Fe+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O |
Fe2H2O13S3 | |
MFCD00011007 | |
167265 |
Specifications
15244-10-7 | |
Granules, Powder | |
Fe2H2O13S3 | |
Soluble in waterSoluble in water. Sparingly soluble in alcohol. | |
O.[Fe+3].[Fe+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O | |
417.87 | |
399.87 (Anhydrous) | |
Odorless | |
Grayish-white powder or granules | |
Iron(III) sulfate hydrate |
Gray-White | |
250 g | |
MFCD00011007 | |
VXWSFRMTBJZULV-UHFFFAOYSA-H | |
diiron(3+) hydrate trisulfate | |
167265 | |
Reagent | |
Hygroscopic | |
2.0-2.1 |
Safety and Handling
P201-P202-P264b-P270-P281-P301+P312-P302+P352-P305+P351+P338-P308+P313-P310-P330-P332+P313-P362-P501c
H302-H315-H318-H350
DOTInformation : Hazard Class: 8; Packaging Group: III
EINECSNumber : 233-072-9
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only