Learn More
Iron(III) trifluoromethanesulfonate, tech. 90%, Thermo Scientific Chemicals
$42.92 - $392.63
Chemical Identifiers
CAS | 63295-48-7 |
---|---|
Molecular Formula | C3F9FeO9S3 |
Molecular Weight (g/mol) | 503.035 |
MDL Number | MFCD15144785 |
InChI Key | OSHOQERNFGVVRH-UHFFFAOYSA-K |
Synonym | iron iii trifluoromethanesulfonate, fe otf 3, ferric triflate, iron iii triflate, fe cf3so3 3, ksc497q0p, iron 3+ tritriflate, ferric trifluoromethanesulfonate, iron 1,1,1-trifluoromethanesulfonic acid, iron 3+ ion tritrifluoromethanesulfonate |
PubChem CID | 11656316 |
IUPAC Name | iron(3+);trifluoromethanesulfonate |
SMILES | C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Fe+3] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAH60034MD
|
Thermo Scientific Chemicals
H60034MD |
250 mg |
Each for $42.92
|
|
||||||
AAH6003403
|
Thermo Scientific Chemicals
H6003403 |
1 g |
Each for $100.07
|
|
||||||
AAH6003406
|
Thermo Scientific Chemicals
H6003406 |
5 g |
Each for $392.63
|
|
||||||
Description
Iron(III) triflate is used as an efficient catalyst in the synthesis of 2,3-unsaturated-O-glycosides from 2,4,6-tri-O-acetyl-D-glucal. It is also involved in the synthesis of a variety of beta-enamino ketones and esters.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIron(III) triflate is used as an efficient catalyst in the synthesis of 2,3-unsaturated-O-glycosides from 2,4,6-tri-O-acetyl-D-glucal. It is also involved in the synthesis of a variety of beta-enamino ketones and esters.
Solubility
Soluble in water, methanol and acetonitrile.
Notes
Moisture sensitive. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
Chemical Identifiers
63295-48-7 | |
503.035 | |
OSHOQERNFGVVRH-UHFFFAOYSA-K | |
11656316 | |
C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Fe+3] |
C3F9FeO9S3 | |
MFCD15144785 | |
iron iii trifluoromethanesulfonate, fe otf 3, ferric triflate, iron iii triflate, fe cf3so3 3, ksc497q0p, iron 3+ tritriflate, ferric trifluoromethanesulfonate, iron 1,1,1-trifluoromethanesulfonic acid, iron 3+ ion tritrifluoromethanesulfonate | |
iron(3+);trifluoromethanesulfonate |
Specifications
63295-48-7 | |
110°C (230°F) | |
Fe(OSO2CF3)3 | |
250 mg | |
iron iii trifluoromethanesulfonate, fe otf 3, ferric triflate, iron iii triflate, fe cf3so3 3, ksc497q0p, iron 3+ tritriflate, ferric trifluoromethanesulfonate, iron 1,1,1-trifluoromethanesulfonic acid, iron 3+ ion tritrifluoromethanesulfonate | |
OSHOQERNFGVVRH-UHFFFAOYSA-K | |
iron(3+);trifluoromethanesulfonate | |
11656316 | |
90% | |
Iron(III) trifluoromethanesulfonate |
183°C | |
C3F9FeO9S3 | |
MFCD15144785 | |
Moisture sensitive | |
Soluble in water,methanol and acetonitrile. | |
C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Fe+3] | |
503.035 | |
503.04 | |
Technical |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only