missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Iron nitride, Thermo Scientific Chemicals
$363.00 - $1281.78
Chemical Identifiers
CAS | 37245-77-5 |
---|---|
Molecular Formula | Fe6H2N2-2 |
Molecular Weight (g/mol) | 365.1 |
MDL Number | MFCD00134717 |
InChI Key | ILMFDGFXPJCFQW-UHFFFAOYSA-N |
Synonym | iron nitride,-325 mesh powder, 1-aza-2,3,4-triferracyclobutan-1-yliron; diferrioamine |
PubChem CID | 56843031 |
IUPAC Name | azanide;azanidylideneiron;iron |
SMILES | [NH2-].[N-]=[Fe].[Fe].[Fe].[Fe].[Fe].[Fe] |
Description
Iron nitride is used in the synthesis of ammonia and carbon nanofibers. It is involved in the creation of ferrofluids, a liquid rocket fuel. It finds applications in making the strongest naturally magnetic material.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
37245-77-5 | |
365.1 | |
ILMFDGFXPJCFQW-UHFFFAOYSA-N | |
56843031 | |
[NH2-].[N-]=[Fe].[Fe].[Fe].[Fe].[Fe].[Fe] |
Fe6H2N2-2 | |
MFCD00134717 | |
iron nitride,-325 mesh powder, 1-aza-2,3,4-triferracyclobutan-1-yliron; diferrioamine | |
azanide;azanidylideneiron;iron |
Specifications
Gray | |
Powder | |
Fe6H2N2-2 | |
iron nitride,-325 mesh powder, 1-aza-2,3,4-triferracyclobutan-1-yliron; diferrioamine | |
ILMFDGFXPJCFQW-UHFFFAOYSA-N | |
azanide;azanidylideneiron;iron | |
56843031 | |
Gray powder |
37245-77-5 | |
25 g | |
MFCD00134717 | |
Insoluble in water. | |
[NH2-].[N-]=[Fe].[Fe].[Fe].[Fe].[Fe].[Fe] | |
365.1 | |
Odorless | |
Iron nitride |
Safety and Handling
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only