missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Iron powder, spherical, 1-3 micron, 98+%, Thermo Scientific Chemicals

Carbonyl iron powder

Supplier:  Thermo Scientific Chemicals 04033722

 View more versions of this product

Catalog No. AA4033722


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Gray
7439-89-6
>98% (Metals basis)
MFCD00010999
ferrum, iron, elemental, ferrous, remko, armco, ferrovac, hoeganaes eh, 3zhp, ancor b, atomiron 5m
[Fe]
55.85
CHEBI:82664
Iron
Gray powder
Fe
100 g
XEEYBQQBJWHFJM-UHFFFAOYSA-N
iron
23925
Powder

Chemical Identifiers

7439-89-6
55.85
XEEYBQQBJWHFJM-UHFFFAOYSA-N
23925
iron
Fe
MFCD00010999
ferrum, iron, elemental, ferrous, remko, armco, ferrovac, hoeganaes eh, 3zhp, ancor b, atomiron 5m
CHEBI:82664
[Fe]
Videos
Safety and Handling

Safety and Handling

P235+P410-P280

H500-X

DOTInformation : DOT Class: 4.2; Packaging Group: II

EINECSNumber : 231-096-4

RTECSNumber : NO4565500

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only