missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Iron sponge, -100 mesh, 99.9% (metals basis), Thermo Scientific Chemicals
$63.00 - $325.05
Chemical Identifiers
CAS | 7439-89-6 |
---|---|
Molecular Formula | Fe |
Molecular Weight (g/mol) | 55.85 |
MDL Number | MFCD00010999 |
InChI Key | XEEYBQQBJWHFJM-UHFFFAOYSA-N |
Synonym | ferrum, iron, elemental, ferrous, remko, armco, ferrovac, hoeganaes eh, 3zhp, ancor b, atomiron 5m |
PubChem CID | 23925 |
ChEBI | CHEBI:82664 |
IUPAC Name | iron |
SMILES | [Fe] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA4324022
|
Thermo Scientific Chemicals
04324022 |
100 g |
Each for $63.00
|
|
|||||
AA4324036
|
Thermo Scientific Chemicals
04324036 |
500 g |
Each for $120.53
|
|
|||||
AA43240A3
|
Thermo Scientific Chemicals
043240A3 |
2 kg |
Each for $325.05
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7439-89-6 | |
55.85 | |
XEEYBQQBJWHFJM-UHFFFAOYSA-N | |
23925 | |
iron |
Fe | |
MFCD00010999 | |
ferrum, iron, elemental, ferrous, remko, armco, ferrovac, hoeganaes eh, 3zhp, ancor b, atomiron 5m | |
CHEBI:82664 | |
[Fe] |
Specifications
Gray | |
Fe | |
UN3089 | |
ferrum, iron, elemental, ferrous, remko, armco, ferrovac, hoeganaes eh, 3zhp, ancor b, atomiron 5m | |
[Fe] | |
55.85 | |
CHEBI:82664 | |
−100 Mesh | |
Iron sponge |
7439-89-6 | |
MFCD00010999 | |
100 g | |
XEEYBQQBJWHFJM-UHFFFAOYSA-N | |
iron | |
23925 | |
99.9% | |
Sponge | |
(metals basis) |
Safety and Handling
GHS H Statement
H228-H319-H335
Flammable solid.
Causes serious eye irritation.
May cause respiratory irritation.
P235+P410-P280
H500-X
DOTInformation : Hazard Class: 4.2; Packaging Group: II
EINECSNumber : 231-096-4
RTECSNumber : NO4565500
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only