missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Iron sputtering target, 76.2mm (3.0 in.) dia. x 6.35mm (0.250 in.) thick, 99.95% (metals basis), Thermo Scientific™
Supplier: Thermo Scientific Chemicals 40857KS
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Iron sputtering target | |
7439-89-6 | |
Fe | |
1 Ea. | |
XEEYBQQBJWHFJM-UHFFFAOYSA-N | |
iron | |
23925 | |
99.95% |
76.2mm (3.0 in.) dia. x 6.35mm (0.250 in.) thick | |
(metals basis) | |
MFCD00010999 | |
ferrum, iron, elemental, ferrous, remko, armco, ferrovac, hoeganaes eh, 3zhp, ancor b, atomiron 5m | |
[Fe] | |
55.85 | |
CHEBI:82664 |
Chemical Identifiers
7439-89-6 | |
55.85 | |
XEEYBQQBJWHFJM-UHFFFAOYSA-N | |
23925 | |
iron |
Fe | |
MFCD00010999 | |
ferrum, iron, elemental, ferrous, remko, armco, ferrovac, hoeganaes eh, 3zhp, ancor b, atomiron 5m | |
CHEBI:82664 | |
[Fe] |
Safety and Handling
H500
EINECSNumber : 231-096-4
TSCA : Yes
Recommended Storage : Ambient temperatures