missing translation for 'onlineSavingsMsg'
Learn More

Iron standard solution, for AAS, 1 mg/ml Fe in 2 to 5% HNO3

CAS: 7439-89-6 | Fe | 55.85 g/mol

Supplier:  Thermo Scientific Chemicals 196055000

 View more versions of this product

Catalog No. AC196055000


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For atomic adsorption spectroscopy
Specifications

Specifications

Iron standard solution
Plastic bottle
7.86
Fe
MFCD00010999
Miscible with water
[Fe]
55.85
23925
55.85
1 mg/ml Fe in 2% HNO3
7439-89-6,7697-37-2,7732-18-5
Liquid
997 to 1003 μg/mL of Fe (3+) in 2% HNO3 (20°C)
Colorless
XEEYBQQBJWHFJM-UHFFFAOYSA-N
iron
500 mL
CHEBI:82664
AAS
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only