missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Iron wire, 0.5mm (0.02in) dia, Puratronic™, 99.995% (metals basis), Thermo Scientific Chemicals

$337.85 - $1117.62

Chemical Identifiers

CAS 7439-89-6
Molecular Formula Fe
Molecular Weight (g/mol) 55.85
MDL Number MFCD00010999
InChI Key XEEYBQQBJWHFJM-UHFFFAOYSA-N
Synonym ferrum, iron, elemental, ferrous, remko, armco, ferrovac, hoeganaes eh, 3zhp, ancor b, atomiron 5m
PubChem CID 23925
ChEBI CHEBI:82664
IUPAC Name iron
SMILES [Fe]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA10936G1
View Documents
Thermo Scientific Chemicals
010936G1
1 m
Each for $337.85
 
AA10936G5
View Documents
Thermo Scientific Chemicals
010936G5
5 m
Each for $1,117.62
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7439-89-6
55.85
XEEYBQQBJWHFJM-UHFFFAOYSA-N
23925
iron
Fe
MFCD00010999
ferrum, iron, elemental, ferrous, remko, armco, ferrovac, hoeganaes eh, 3zhp, ancor b, atomiron 5m
CHEBI:82664
[Fe]

Specifications

7439-89-6
MFCD00010999
ferrum, iron, elemental, ferrous, remko, armco, ferrovac, hoeganaes eh, 3zhp, ancor b, atomiron 5m
[Fe]
55.85
CHEBI:82664
Puratronic™
Wire
(metals basis)
Fe
1 m
XEEYBQQBJWHFJM-UHFFFAOYSA-N
iron
23925
99.995%
≈1.5g/m
Iron wire, 0.5mm (0.02 in.) dia.
Videos
Safety and Handling

Safety and Handling

H500

EINECSNumber : 231-096-4

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only