Air Boss™ Defense IRT Water Storage Bladders DFS Item
Effectively collect wastewater runoff from IRT decontamination shelters
Supplier:  Air Boss™ Defense WSB250G
 View more versions of this product
Catalog No. 19-130-0635
Explore available promotions
Includes: Protective ground cloth and transport bag
Description
Description
  • Co-polymer material is highly resistant to chemicals, mold and mildew
  • Integral pressure relief valve, ball valve, back flow prevention valve, and cam-lok
  • Ball valve on either side of cam-lok for shut off, backflow prevention for one way fluid exchange, cam-lok for quick disconnect
  • Rugged design allows for use in the toughest environments
  • Quickly and easily deploys, no tools required, reusable and repairable
  • Compact and lightweight for portability, fully collapsible and foldable
Specifications
Specifications
Waste Storage Bladder
250 gal.
Collects waste water run-off from IRT decontamination shelters
Polyester
346.3 L
Protective ground Cloth and transport bag, integral pressure relief valve, ball valve, back flow prevention valve and cam-lok