Learn More
Isethionic acid sodium salt, 98%
CAS: 1562-00-1 | C2H5NaO4S | 148.108 g/mol
$128.70 - $451.92
Chemical Identifiers
| CAS | 1562-00-1 |
|---|---|
| Molecular Formula | C2H5NaO4S |
| Molecular Weight (g/mol) | 148.108 |
| MDL Number | MFCD00007534 |
| InChI Key | LADXKQRVAFSPTR-UHFFFAOYSA-M |
| Synonym | sodium 2-hydroxyethanesulfonate, sodium isethionate, isethionic acid sodium salt, 2-hydroxyethanesulfonic acid sodium salt, sodium hydroxyethylsulfonate, ethanesulfonic acid, 2-hydroxy-, monosodium salt, sodium 2-hydroxyethylsulfonate, sodium 2-hydroxyethanesulphonate, sodium beta-hydroxyethanesulfonate, sodium 1-hydroxy-2-ethanesulfonate |
| PubChem CID | 517063 |
| IUPAC Name | sodium;2-hydroxyethanesulfonate |
| SMILES | C(CS(=O)(=O)[O-])O.[Na+] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA120540B
|
Thermo Scientific Chemicals
A120540B |
1000 g |
Each for $128.70
|
|
|||||
|
AAA120540I
|
Thermo Scientific Chemicals
A120540I |
5000 g |
Each for $451.92
|
|
|||||
Description
Isethionic acid sodium salt is an amphoteric detergent used in detergent bar soaps. It makes a dense lather in addition to the lather made by the soap. It is mild on the skin, and non-drying. It works equally well in soft or hard water. It is also an anti-static agent in shampoos.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIsethionic acid sodium salt is an amphoteric detergent used in detergent bar soaps. It makes a dense lather in addition to the lather made by the soap. It is mild on the skin, and non-drying. It works equally well in soft or hard water. It is also an anti-static agent in shampoos.
Solubility
Soluble in water.
Notes
Store in cool place. Keep away from oxidizing agents.
Chemical Identifiers
| 1562-00-1 | |
| 148.108 | |
| LADXKQRVAFSPTR-UHFFFAOYSA-M | |
| 517063 | |
| C(CS(=O)(=O)[O-])O.[Na+] |
| C2H5NaO4S | |
| MFCD00007534 | |
| sodium 2-hydroxyethanesulfonate, sodium isethionate, isethionic acid sodium salt, 2-hydroxyethanesulfonic acid sodium salt, sodium hydroxyethylsulfonate, ethanesulfonic acid, 2-hydroxy-, monosodium salt, sodium 2-hydroxyethylsulfonate, sodium 2-hydroxyethanesulphonate, sodium beta-hydroxyethanesulfonate, sodium 1-hydroxy-2-ethanesulfonate | |
| sodium;2-hydroxyethanesulfonate |
Specifications
| 1562-00-1 | |
| C2H5NaO4S | |
| 1000 g | |
| sodium 2-hydroxyethanesulfonate, sodium isethionate, isethionic acid sodium salt, 2-hydroxyethanesulfonic acid sodium salt, sodium hydroxyethylsulfonate, ethanesulfonic acid, 2-hydroxy-, monosodium salt, sodium 2-hydroxyethylsulfonate, sodium 2-hydroxyethanesulphonate, sodium beta-hydroxyethanesulfonate, sodium 1-hydroxy-2-ethanesulfonate | |
| LADXKQRVAFSPTR-UHFFFAOYSA-M | |
| sodium;2-hydroxyethanesulfonate | |
| 517063 | |
| 98% |
| 191°C to 194°C | |
| MFCD00007534 | |
| 3633992 | |
| Soluble in water. | |
| C(CS(=O)(=O)[O-])O.[Na+] | |
| 148.108 | |
| 148.11 | |
| Isethionic acid sodium salt |
Safety and Handling
EINECSNumber : 216-343-6
RTECSNumber : KI7700000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only